Novus Biologicals
Manufacturer Code:NBP287477
Catalog # NB035099
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human G protein alpha-13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G alpha-13; G alpha-13 G13 G-protein subunit alpha-13 guanine nucleotide binding protein (G protein) alpha 13 guanine nucleotide-binding protein subunit alpha-13 MGC46138; G-protein subunit alpha-13; guanine nucleotide binding protein (G protein), alpha 13; Guanine nucleotide-binding protein subunit alpha-13
Gene Aliases: G13; GNA13
UniProt ID: (Human) F5H1G8
Entrez Gene ID: (Human) 10672
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.