Novus Biologicals
Manufacturer Code:NBP302979100UL
Catalog # NB092884
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 250-350 of human G Protein alpha z (NP_002064.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G(x) alpha chain; G(x) alpha chain guanine nucleotide binding protein (G protein) alpha z polypeptide guanine nucleotide-binding protein G(z) subunit alpha gz-alpha transducin alpha; guanine nucleotide binding protein (G protein), alpha z polypeptide; Guanine nucleotide-binding protein G(z) subunit alpha; Gz-alpha; transducin alpha
Gene Aliases: GNAZ
UniProt ID: (Human) P19086
Entrez Gene ID: (Human) 2781
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.