Novus Biologicals
Manufacturer Code:NB500517
Catalog # NB500517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | FYN-01 |
Immunogen | Bacterial expressed recombinant fragment corresponding to amino acids 7-176 of Human Fyn. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-syn protooncogene; EC 2.7.10 EC 2.7.10.2 FYN oncogene related to SRC FGR YES MGC45350 OKT3-induced calcium influx regulator p59-Fyn protein-tyrosine kinase fyn Proto-oncogene c-Fyn Proto-oncogene Syn proto-oncogene tyrosine-protein kinase fyn SLKc-syn protooncogene src/yes-related novel Src-like kinase SYN tyrosine kinase p59fyn(T) tyrosine-protein kinase Fyn; FYN oncogene related to SRC, FGR, YES; OKT3-induced calcium influx regulator; p59-Fyn; Proto-oncogene c-Fyn; Proto-oncogene Syn; SLK; Src-like kinase; src/yes-related novel; tyrosine kinase p59fyn(T); Tyrosine-protein kinase Fyn
Gene Aliases: FYN; p59-FYN; SLK; SYN
UniProt ID: (Human) P06241
Entrez Gene ID: (Human) 2534
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.