Invitrogen
Manufacturer Code:PA19045
Catalog # 01-674-629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the N terminus amino acids VQCKDKEATKLTE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: c-syn protooncogene; FYN oncogene related to SRC, FGR, YES; MGC45350 SLK SYN; OKT3-induced calcium influx regulator; p59-Fyn; Proto-oncogene c-Fyn; Proto-oncogene Syn; proto-oncogene tyrosine-protein kinase Fyn; SLK; Src Kinase p59; Src-like kinase; src/yes-related novel; tyrosine kinase p59fyn(T); Tyrosine-protein kinase Fyn
Gene Aliases: AI448320; AW552119; FYN; p59-FYN; SLK; SYN
UniProt ID: (Human) P06241, (Mouse) P39688
Entrez Gene ID: (Human) 2534, (Mouse) 14360
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.