Abnova
Manufacturer Code:MAB0116
Catalog # 89-107-882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 41AT858.235 |
Immunogen | Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S68 of mouse Fxyd1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic phosphopeptide of Fxyd1.
For Research Use Only
Protein Aliases: 0610012C17Rik; 1110006M24Rik; FXYD domain-containing ion transport regulator 1; Phospholemman; PLM; PML; Sodium/potassium-transporting ATPase subunit FXYD1
Gene Aliases: 0610012C17Rik; 1110006M24Rik; Fxyd1; Plm; Pml
UniProt ID: (Mouse) Q9Z239
Entrez Gene ID: (Mouse) 56188
Molecular Function:
enzyme modulator
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.