Abnova
Manufacturer Code:MAB2413
Catalog # 89-110-995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MC-480 |
Immunogen | Native purified from F9 tetracarcinoma stem cells (X-irradiated). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for short term. For long term storage store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native Fut4.
For Research Use Only
Protein Aliases: 3-fucosyl-N-acetyl-lactosamine epitope; AI451562; alpha (1,3) fucosyltransferase, myeloid specific; alpha 1,3-fucosyltransferase; Alpha-(1,3)-fucosyltransferase 4; FAL; Fuc-TIV; Fucosyltransferase 4; Fucosyltransferase IV; FucT-IV; Galactoside 3-L-fucosyltransferase; Lewis x; LeX; stage-specific embryonic antigen 1
Gene Aliases: AI451562; CD15; Elft; FAL; FucT-IV; Fut4; LeX; SSEA-1; Ssea1
UniProt ID: (Mouse) Q11127
Entrez Gene ID: (Mouse) 14345
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.