Novus Biologicals
Manufacturer Code:NBP212321AF750
Catalog # NP212321A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0298-4G31 |
Immunogen | Human Furin |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: dibasic processing enzyme; Dibasic-processing enzyme; Dibasic-processing enzyme EC 3.4.21 EC 3.4.21.75 FURdibasic processing enzyme furin (paired basic amino acid cleaving enzyme) furin membrane associated receptor protein PACEFES upstream region paired basic amino acid cleaving enzyme (furin membrane associated receptorprotein) Paired basic amino acid residue-cleaving enzyme PCSK3furin proprotein convertase subtilisin/kexin type 3 SPC1; FES upstream region; Furin; furin (paired basic amino acid cleaving enzyme); furin, membrane associated receptor protein; PACE; Paired basic amino acid residue-cleaving enzyme; proprotein convertase subtilisin/kexin type 3
Gene Aliases: FUR; FURIN; PACE; PCSK3; SPC1
UniProt ID: (Human) P09958
Entrez Gene ID: (Human) 5045
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.