R&D Systems
Manufacturer Code:IC1503T100UG
Catalog # IC1503T100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 222722 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Furin Asp108-Glu715 Accession # NP_002560 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: dibasic processing enzyme; Dibasic-processing enzyme; Dibasic-processing enzyme EC 3.4.21 EC 3.4.21.75 FUR FURdibasic processing enzyme furin (paired basic amino acid cleaving enzyme) furin membrane associated receptor protein PACE PACEFES upstream region paired basic amino acid cleaving enzyme (furin membrane associated receptorprotein) Paired basic amino acid residue-cleaving enzyme PC PCSK3 PCSK3furin proprotein convertase subtilisin/kexin type 3 SPC1; FES upstream region; Furin; furin (paired basic amino acid cleaving enzyme); furin, membrane associated receptor protein; PACE; Paired basic amino acid residue-cleaving enzyme; proprotein convertase subtilisin/kexin type 3
Gene Aliases: FUR; FURIN; PACE; PCSK3; SPC1
UniProt ID: (Human) P09958
Entrez Gene ID: (Human) 5045
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.