Abnova
Manufacturer Code:PAB8731
Catalog # 89-133-888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 779 of mouse Furin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Furin.
For Research Use Only
Protein Aliases: 9130404I01Rik; Dibasic-processing enzyme; Fur; Furin; PACE; paired basic amino acid residue cleaving enzyme; Paired basic amino acid residue-cleaving enzyme; Pcsk3; Prohormone convertase 3; proprotein convertase subtilisin/kexin type 3; SPC1; subtilisin pro-protein processing enzyme
Gene Aliases: 9130404I01Rik; Fur; Furin; PACE; Pcsk3; SPC1
UniProt ID: (Mouse) P23188
Entrez Gene ID: (Mouse) 18550
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.