R&D Systems
Manufacturer Code:AF5768SP
Catalog # AF5768SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Fucosyltransferase 8/FUT8 Asp32-Lys575 Accession # Q9BYC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 16 Fucosyltransferase alpha-(16)-fucosyltransferase alpha1-6FucT EC 2.4.1.68 Fucosyltransferase 8 fucosyltransferase 8 (alpha (16) fucosyltransferase) FUT8 GDP-fucose--glycoprotein fucosyltransferase GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha16-fucosyltransferase Glycoprotein 6-alpha-L-fucosyltransferase MGC26465; Alpha-(1,6)-fucosyltransferase; Alpha1-6FucT; Fucosyltransferase 8; fucosyltransferase 8 (alpha (1,6) fucosyltransferase); GDP-fucose--glycoprotein fucosyltransferase; GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1,6-fucosyltransferase; Glycoprotein 6-alpha-L-fucosyltransferase
Gene Aliases: FUT8
UniProt ID: (Human) Q9BYC5
Entrez Gene ID: (Human) 2530
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.