Novus Biologicals
Manufacturer Code:NBP302941100UL
Catalog # NB092872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Fucosyltransferase 7/FUT7 (NP_004470.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-(1,3)-fucosyltransferase 7; alpha-(13)-fucosyltransferase EC 2.4.1 EC 2.4.1.- EC 2.4.1.65 Fucosyltransferase 7 fucosyltransferase 7 (alpha (13) fucosyltransferase) Fucosyltransferase VII fuc-TVII FucT-VII Galactoside 3-L-fucosyltransferase Selectin ligand synthase selectin-ligand synthase; Fuc-TVII; Fucosyltransferase 7; fucosyltransferase 7 (alpha (1,3) fucosyltransferase); Fucosyltransferase VII; Galactoside 3-L-fucosyltransferase; Selectin ligand synthase; selectin-ligand synthase
Gene Aliases: FucT-VII; FUT7
UniProt ID: (Human) Q11130
Entrez Gene ID: (Human) 2529
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.