Novus Biologicals
Manufacturer Code:NBP300239H
Catalog # NB065025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Fucose Mutarotase (A2VDF0-1 Met1-Leu154). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C10orf125 chromosome 10 open reading frame 125 FucM fucose mutarotase FUCU; Fucose mutarotase; protein fucU homolog
Gene Aliases: C10orf125; FucM; FUCU; FUOM
UniProt ID: (Human) A2VDF0
Entrez Gene ID: (Human) 282969
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.