R&D Systems
Manufacturer Code:MAB194SP
Catalog # MAB194SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 145901 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-4 Phe37-Glu180 Accession # Q61088 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD344; CD344 CD344 antigen EVR1 exudative vitreoretinopathy 1 FEVR frizzled (Drosophila) homolog 4 frizzled homolog 4 (Drosophila) frizzled-4 Frizzled4 Fz-4 FZD4 FZD4S FzE4 GPCR hFz4 MGC34390 WNT receptor frizzled-4; frizzled 4, seven transmembrane spanning receptor; frizzled family receptor 4; frizzled homolog 4; Frizzled-4; Fz-4; FzE4; WNT receptor frizzled-4
Gene Aliases: CD344; EVR1; FEVR; Fz-4; Fz4; FZD4; FZD4S; FzE4; GPCR; hFz4
UniProt ID: (Human) Q9ULV1
Entrez Gene ID: (Human) 8322
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.