R&D Systems
Manufacturer Code:FAB1001S100UG
Catalog # FAB1001S100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 169310 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-3 Met1-Val157 Accession # Q61086 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: frizzled (Drosophila) homolog 3 frizzled homolog 3 (Drosophila) frizzled-3 Frizzled3 Fz-3 FZD3 hFz3; frizzled 3, seven transmembrane spanning receptor; frizzled family receptor 3; frizzled homolog 3; Frizzled-3; Fz-3
Gene Aliases: Fz-3; FZD3
UniProt ID: (Human) Q9NPG1
Entrez Gene ID: (Human) 7976
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.