Invitrogen
Manufacturer Code:PA533543
Catalog # PIPA533543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from 3rd extracellular domain of human Frizzled 2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: frizzled 2, seven transmembrane spanning receptor; frizzled family receptor 2; frizzled homolog 2; Frizzled-2; Fz-2; FZD2 Dfrizzled2 Fz2 Frizzled family receptor 2 Frizzled-2 HFz2 Frizzled homolog 2 Fz-2 FzE2 Mfz10 Rfz2 Frizzled 2; FzE2
Gene Aliases: fz-2; Fz2; FZD2; fzE2; hFz2
UniProt ID: (Human) Q14332
Entrez Gene ID: (Human) 2535
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.