Novus Biologicals
Manufacturer Code:NBP120920
Catalog # NBP120920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence RYATLEHPFHCPR corresponding to internal region according to NP_001457.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: frizzled (Drosophila) homolog 2 frizzled homolog 2 (Drosophila) frizzled-2 fz-2 Fz2 FzE2 hFz2; frizzled 2, seven transmembrane spanning receptor; frizzled family receptor 2; frizzled homolog 2; Frizzled-2; Fz-2; FzE2
Gene Aliases: fz-2; Fz2; FZD2; fzE2; hFz2
UniProt ID: (Human) Q14332
Entrez Gene ID: (Human) 2535
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.