Proteintech
Manufacturer Code:242721AP150UL
Catalog # 50-173-4269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Frizzled 2 Fusion Protein Ag18003 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 2; Frizzled 2 Fz 2 FZD2 FzE2 hFz2; frizzled 2, seven transmembrane spanning receptor; frizzled family receptor 2; frizzled homolog 2; Frizzled-10; Frizzled-2; Fz-10; Fz-2; FzE2; mFz2; rFz2
Gene Aliases: AL033370; AW456835; Fz-10; fz-2; Fz10; Fz2; Fzd10; FZD2; fzE2; hFz2; Mfz10; Mfz10a
UniProt ID: (Human) Q14332, (Rat) Q08464, (Mouse) Q9JIP6
Entrez Gene ID: (Human) 2535, (Rat) 64512, (Mouse) 57265
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.