Novus Biologicals
Manufacturer Code:NBP212316C
Catalog # NBP212316C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0235-9A12 |
Immunogen | Mouse Frizzled-1 |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: DKFZp564G072 FLJ95923 frizzled family receptor 1 frizzled homolog 1 frizzled Drosophila homolog of 1 frizzled-1 fz-1 fzE1 hFz1 seven transmembrane spanning receptor Wnt receptor; frizzled 1, seven transmembrane spanning receptor; frizzled family receptor 1; frizzled homolog 1; frizzled, Drosophila, homolog of, 1; Frizzled-1; Fz-1; FzE1; hFz1; Wnt receptor
Gene Aliases: FZD1
UniProt ID: (Human) Q9UP38
Entrez Gene ID: (Human) 8321
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.