R&D Systems
Manufacturer Code:MAB11201SP
Catalog # MAB11201SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 162531 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-1 Gln72-His248 (Met122Ile) Accession # O70421 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp564G072 FLJ95923 frizzled family receptor 1 frizzled homolog 1 frizzled Drosophila homolog of 1 frizzled-1 Frizzled1 fz-1 FZD1 fzE1 hFz1 seven transmembrane spanning receptor Wnt receptor; frizzled 1, seven transmembrane spanning receptor; frizzled family receptor 1; frizzled homolog 1; frizzled, Drosophila, homolog of, 1; Frizzled-1; Fz-1; FzE1; hFz1; Wnt receptor
Gene Aliases: FZD1
UniProt ID: (Human) Q9UP38
Entrez Gene ID: (Human) 8321
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.