Novus Biologicals
Manufacturer Code:NBP270766AF532
Catalog # NP270766A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C12 |
Immunogen | Full length recombinant protein of human Frataxin (NP_000135) produced in HEK293T cells. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CyaY EC 1.16.3.1 FA FARRmitochondrial frataxin Friedreich ataxia Friedreich ataxia protein Fxn X25MGC57199; d-FXN; Frataxin intermediate form; Frataxin mature form; Frataxin(56-210); Frataxin(78-210); Frataxin(81-210); Frataxin, mitochondrial; Friedreich ataxia protein; Fxn; i-FXN; m56-FXN; m78-FXN; m81-FXN
Gene Aliases: CyaY; FA; FARR; FRDA; FXN; X25
UniProt ID: (Human) Q16595
Entrez Gene ID: (Human) 2395
Molecular Function: cation transporter kinase transferase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.