Abnova
Manufacturer Code:PAB8724
Catalog # 89-133-882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 55 of mouse Foxh1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Foxh1.
For Research Use Only
Protein Aliases: Fast-1; Fast-2; FAST1 transcription factor; Fast2; Forkhead activin signal transducer 1; Forkhead activin signal transducer 2; Forkhead box protein H1
Gene Aliases: fast-1; fast-2; Fast1; Fast2; Foxh1
UniProt ID: (Mouse) O88621
Entrez Gene ID: (Mouse) 14106
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.