Novus Biologicals
Manufacturer Code:NBP233297
Catalog # NBP233297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G3 |
Immunogen | Full-length His-tagged murine FOXP3 was used as the immunogen. The precise epitope is not known but it has been mapped to the N-terminal portion of the protein (NP_001107849). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AIID AIIDMGC141961 DIETER forkhead box P3 Forkhead Box Protein P3 FOXP3delta7 immune dysregulation polyendocrinopathy enteropathy X-linked Immunodeficiency Polyendocrinopathy Enteropathy X-Linked IPEX JM2 MGC141961 MGC141963 PIDX PIDXMGC141963 Scurfin XPID XPIDpolyendocrinopathy enteropathy X-linked; Forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; Scurfin
Gene Aliases: AIID; DIETER; FOXP3; IPEX; JM2; PIDX; XPID
UniProt ID: (Human) Q9BZS1
Entrez Gene ID: (Human) 50943
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.