Novus Biologicals
Manufacturer Code:NBP186672
Catalog # NBP186672
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SLGYGAALNASLQAALAESSLPLLSNPGLINNASSGLLQAVHEDLNGSLDHIDSNGNSSPGCSPQPHIHSIHVKEEPVIAEDEDCPMSLVTTANHSPELEDDRE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAG repeat protein 44; CAG repeat protein 44 CAGH44TNRC10forkhead box protein P2 DKFZp686H1726 forkhead box P2 forkhead/winged-helix transcription factor SPCH1 trinucleotide repeat containing 10 Trinucleotide repeat-containing gene 10 protein; Forkhead box protein P2; forkhead/winged-helix transcription factor; trinucleotide repeat containing 10; Trinucleotide repeat-containing gene 10 protein
Gene Aliases: CAGH44; FOXP2; SPCH1; TNRC10
UniProt ID: (Human) O15409
Entrez Gene ID: (Human) 93986
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.