R&D Systems
Manufacturer Code:MAB45341
Catalog # MAB45341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 837016 |
Immunogen | <EM>E. coli</EM>-derived recombinant human FoxP1 Lys548-Glu677 Accession # Q9H334 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ23741 fork head-related protein like B forkhead box P1 forkhead box protein P1 glutamine-rich factor 1 hFKH1B HSPC215 MGC12942 MGC88572 MGC99551 QRF1 QRF112CC4; fork head-related protein like B; Forkhead box protein P1; glutamine-rich factor 1; Mac-1-regulated forkhead; MFH
Gene Aliases: 12CC4; FOXP1; hFKH1B; HSPC215; MFH; QRF1
UniProt ID: (Human) Q9H334
Entrez Gene ID: (Human) 27086
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.