Novus Biologicals
Manufacturer Code:NBP237450
Catalog # NBP237450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 60000 |
Immunogen | Purified recombinant fragment of human FOXP1 (AA: 481-677) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ23741 fork head-related protein like B forkhead box P1 forkhead box protein P1 glutamine-rich factor 1 hFKH1B HSPC215 MGC12942 MGC88572 MGC99551 QRF112CC4; fork head-related protein like B; Forkhead box protein P1; glutamine-rich factor 1; Mac-1-regulated forkhead; MFH
Gene Aliases: 12CC4; FOXP1; hFKH1B; HSPC215; MFH; QRF1
UniProt ID: (Human) Q9H334
Entrez Gene ID: (Human) 27086
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.