Novus Biologicals
Manufacturer Code:NBP141091
Catalog # NBP141091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 627 and 677 of human forkhead box P1 using the numbering given in entry NP_116071.2 (GeneID 27086). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
For Research Use Only
Protein Aliases: FLJ23741 fork head-related protein like B forkhead box P1 forkhead box protein P1 glutamine-rich factor 1 hFKH1B HSPC215 MGC12942 MGC88572 MGC99551 QRF112CC4; fork head-related protein like B; Forkhead box protein P1; glutamine-rich factor 1; Mac-1-regulated forkhead; MFH
Gene Aliases: 12CC4; FOXP1; hFKH1B; HSPC215; MFH; QRF1
UniProt ID: (Human) Q9H334
Entrez Gene ID: (Human) 27086
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.