R&D Systems
Manufacturer Code:MAB3619
Catalog # MAB3619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 407003 |
Immunogen | <EM>E. coli</EM>-derived recombinant human FoxJ1 Lys306-Leu421 Accession # Q92949.3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: FKHL13 FKHL13forkhead transcription factor HFH-4 forkhead box J1 forkhead-like 13 Forkhead-related protein FKHL13 Hepatocyte nuclear factor 3 forkhead homolog 4 HFH-4 HFH4fork head homologue 4 HFH-4forkhead box protein J1 MGC35202; fork head homologue 4; Forkhead box protein J1; forkhead transcription factor HFH-4; forkhead-like 13; Forkhead-related protein FKHL13; Hepatocyte nuclear factor 3 forkhead homolog 4; HFH-4
Gene Aliases: FKHL13; FOXJ1; HFH-4; HFH4
UniProt ID: (Human) Q92949
Entrez Gene ID: (Human) 2302
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.