R&D Systems
Manufacturer Code:IC3619V100UG
Catalog # IC3619V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 407003 |
Immunogen | <EM>E. coli</EM>-derived recombinant human FoxJ1 Lys306-Leu421 Accession # Q92949.3 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: FKHL13 FKHL13forkhead transcription factor HFH-4 forkhead box J1 forkhead-like 13 Forkhead-related protein FKHL13 Hepatocyte nuclear factor 3 forkhead homolog 4 HFH-4 HFH4fork head homologue 4 HFH-4forkhead box protein J1 MGC35202; fork head homologue 4; Forkhead box protein J1; forkhead transcription factor HFH-4; forkhead-like 13; Forkhead-related protein FKHL13; Hepatocyte nuclear factor 3 forkhead homolog 4; HFH-4
Gene Aliases: FKHL13; FOXJ1; HFH-4; HFH4
UniProt ID: (Human) Q92949
Entrez Gene ID: (Human) 2302
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.