Novus Biologicals
Manufacturer Code:NBP298870V
Catalog # NB056167
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human FoxF1. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FKHL5MGC105125 forkhead box F1 forkhead box protein F1 Forkhead drosophila homolog-like 5 Forkhead-related activator 1 Forkhead-related protein FKHL5 Forkhead-related transcription factor 1 FREAC-1 FREAC1ACDMPV; Forkhead box protein F1; Forkhead, drosophila, homolog-like 5; Forkhead-related activator 1; Forkhead-related protein FKHL5; Forkhead-related transcription factor 1; FREAC-1
Gene Aliases: ACDMPV; FKHL5; FOXF1; FREAC1
UniProt ID: (Human) Q12946
Entrez Gene ID: (Human) 2294
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.