Novus Biologicals
Manufacturer Code:NBP121715AF647
Catalog # NBPC15AF647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0080-7D11 |
Immunogen | Mouse recombinant FLRG |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: FLRGFSRP follistatin-like 3 (secreted glycoprotein) Follistatin-like protein 3 Follistatin-related gene protein follistatin-related protein 3; follistatin-like 3 (secreted glycoprotein); Follistatin-like protein 3; Follistatin-related gene protein; Follistatin-related protein 3
Gene Aliases: FLRG; FSRP; FSTL3; UNQ674/PRO1308
UniProt ID: (Human) O95633
Entrez Gene ID: (Human) 10272
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.