R&D Systems
Manufacturer Code:AF1288
Catalog # AF1288
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human FLRG Met27-Val263 Accession # O95633 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: FLRGFSRP Follistatin-like 3 follistatin-like 3 (secreted glycoprotein) Follistatin-like protein 3 Follistatin-related gene protein follistatin-related protein 3 FSTL3; follistatin-like 3 (secreted glycoprotein); Follistatin-like protein 3; Follistatin-related gene protein; Follistatin-related protein 3
Gene Aliases: FLRG; FSRP; FSTL3; UNQ674/PRO1308
UniProt ID: (Human) O95633
Entrez Gene ID: (Human) 10272
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.