R&D Systems
Manufacturer Code:AF5245SP
Catalog # AF5245SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Follistatin-like 4/FSTL4 Trp23-Val841 Accession # Q5STE3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Follistatin like 4 follistatin-like 4 Follistatin-like protein 4 follistatin-related protein 4 FSL4 FSTL4 KIAA1061; follistatin-like 4; Follistatin-like protein 4; Follistatin-related protein 4
Gene Aliases: FSTL4; KIAA1061
UniProt ID: (Human) Q6MZW2
Entrez Gene ID: (Human) 23105
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.