Invitrogen
Manufacturer Code:PA523016
Catalog # PIPA523016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | N-terminal partial recombit human FLCN protein expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BHD FLCL BHD skin lesion fibrofolliculoma protein birt-Hogg-Dube syndrome protein; BHD skin lesion fibrofolliculoma protein; Birt-Hogg-Dube syndrome protein; Birt-Hogg-Dube syndrome protein homolog; Folliculin
Gene Aliases: AU014660; B430214A04Rik; BHD; FLCL; FLCN
UniProt ID: (Human) Q8NFG4, (Mouse) Q8QZS3
Entrez Gene ID: (Human) 201163, (Mouse) 216805
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.