Invitrogen
Manufacturer Code:MA512144
Catalog # PIMA512144
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | FSH03 |
Immunogen | Purified human FSH-beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: class 1 follicle-stimulating hormone beta polypeptide; class 2 follicle-stimulating hormone beta polypeptide; follicle stimulating hormone beta subunit; follicle stimulating hormone beta-subunit precursor; follicle stimulating hormone, beta polypeptide; Follicle-stimulating hormone beta subunit; follicule stimulating hormone, beta polypeptide; Follitropin beta chain; Follitropin subunit beta; follitropin, beta chain; FSH-B; FSH-B FSH-beta follicle-stimulating hormone beta subunit follitropin beta chain follitropin subunit beta follitropin beta chain; FSH-beta; prefollicle stimulating hormone beta chain
Gene Aliases: FSHB; HH24
UniProt ID: (Bovine) P04837, (Human) P01225
Entrez Gene ID: (Bovine) 281171, (Human) 2488
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.