Invitrogen
Manufacturer Code:MA514726
Catalog # PIMA514726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Radioimmune Assays (RIA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G6D12 |
Immunogen | Human Follicle Stimulating Hormone |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: follicle stimulating hormone, beta polypeptide; Follicle-stimulating hormone beta subunit; Follitropin beta chain; Follitropin subunit beta; follitropin, beta chain; FSH-alpha LSH-alpha TSH-alpha anterior pituitary glycoprotein hormones common subunit alpha choriogonadotropin alpha chain chorionic gonadotrophin subunit alpha chorionic gonadotropin alpha polypeptide follicle-stimulating hormone alpha chain follicle-stimulating hormone alpha subunit follitropin alpha chain glycoprotein hormones alpha chain luteinizing hormone alpha chain lutropin alpha chain thyroid-stimulating hormone alpha chain thyrotropin alpha chain; FSH-B; FSH-beta
Gene Aliases: FSH; FSHB; Fshbeta; HH24
UniProt ID: (Human) P01225, (Mouse) Q60687
Entrez Gene ID: (Human) 2488, (Mouse) 14308
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.