Invitrogen
Manufacturer Code:PA586814
Catalog # PIPA586814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in Human FSHB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: follicle - stimulating hormone subunit beta; follicle stimulating hormone beta; follicle stimulating hormone, beta polypeptide; Follicle-stimulating hormone beta subunit; follicle-stimulating hormone beta-subunit; follicle-stimulating hormone subunit beta; Follitropin beta chain; Follitropin subunit beta; Follitropin subunit beta Follicle-stimulating hormone beta subunit FSH-B FSH-beta Follitropin beta chain FSHB; follitropin, beta chain; FSH-B; FSH-beta
Gene Aliases: FSH; FSHB; Fshbeta; HH24
UniProt ID: (Human) P01225, (Rat) P18427, (Mouse) Q60687
Entrez Gene ID: (Human) 2488, (Rat) 25447, (Mouse) 14308
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.