Abnova
Manufacturer Code:PAB15091
Catalog # 89-117-437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 800-900 of mouse Flt1. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of Flt1.
For Research Use Only
Protein Aliases: AI323757; Embryonic receptor kinase 2; FLT-1; Fms-like tyrosine kinase 1; sFlt1; Tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; vascular permeability factor receptor; VEGFR-1; VEGFR1
Gene Aliases: AI323757; Emrk2; Flt; Flt-1; Flt1; sFlt1; VEGFR-1; VEGFR1
UniProt ID: (Mouse) P35969
Entrez Gene ID: (Mouse) 14254
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.