R&D Systems
Manufacturer Code:FAB812P
Catalog # FAB812P
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 66903 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Flt-3/Flk-2 Asn27-Asn541 Accession # AAA18947 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD135; CD135 antigen; CD135 CD135 antigen EC 2.7.10 fetal liver kinase 2 FL cytokine receptor Flk-2 FLK2 Flt3 FLT-3 FLT3 receptor tyrosine kinase Fms-like tyrosine kinase 3 fms-related tyrosine kinase 3 growth factor receptor tyrosine kinase type III Stem cell tyrosine kinase 1 STK-1 STK1EC 2.7.10.1 Tyrosine-protein kinase receptor FLT3; fetal liver kinase 2; Fetal liver kinase-2; FL cytokine receptor; FLK-2; FLT-3; Fms-like tyrosine kinase 3; fms-related tyrosine kinase 3; growth factor receptor tyrosine kinase type III; Receptor-type tyrosine-protein kinase FLT3; Stem cell tyrosine kinase 1; STK-1
Gene Aliases: CD135; FLK-2; FLK2; FLT3; STK1
UniProt ID: (Human) P36888
Entrez Gene ID: (Human) 2322
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.