Invitrogen
Manufacturer Code:PA581937
Catalog # PIPA581937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FLOT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ECS-1 ECS1 ESA ESA1 M17S1; Epidermal surface antigen; ESA; Flotillin 2 (epidermal surface antigen 1); Flotillin-2; Membrane component chromosome 17 surface marker 1; Membrane component chromosome 17 surface marker 1 homolog; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1); REG-1; Reggie-1; reggie1-1
Gene Aliases: AI573412; ECS-1; ECS1; ESA; ESA1; FLOT2; M17S1; Reg1; reggie-2
UniProt ID: (Human) Q14254, (Mouse) Q60634, (Rat) Q9Z2S9
Entrez Gene ID: (Human) 2319, (Mouse) 14252, (Rat) 83764
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.