Novus Biologicals
Manufacturer Code:NBP179022AF647
Catalog # NB179022AFX
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 377 and 427 of human Flotillin 1 using the numbering given in entry NP_005794.1 (GeneID 10211). |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: flotillin 1 flotillin-1 integral membrane component of caveolae; Flotillin-1; integral membrane component of caveolae
Gene Aliases: FLOT1
UniProt ID: (Human) O75955
Entrez Gene ID: (Human) 10211
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.