R&D Systems
Manufacturer Code:MAB7278
Catalog # MAB7278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 763103 |
Immunogen | <EM>E. coli</EM>-derived recombinant human CELSR3 Val531-Ser711 Accession # Q9NYQ7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: adhesion G protein-coupled receptor C3; anchor protein; Cadherin EGF LAG seven-pass G-type receptor 3; Cadherin family member 11; Cadherin family member 11 cadherin EGF LAG seven-pass G-type receptor 3 (flamingo homolog Drosophila) CDHF11 CDHF11RESDA1 EGFL1 EGFL1cadherin EGF LAG seven-pass G-type receptor 3 EGF-like protein 1 EGF-like-domain multiple 1 epidermal growth factor-like 1 Epidermal growth factor-like protein 1 Flamingo homolog 1 FMI1anchor protein HFMI1 KIAA0812 MEGF2 MEGF2cadherin EGF LAG seven-pass G-type receptor 3 flamingo (Drosophila) homolog multiple EGF-like domains 2 Multiple EGF-like domains protein 2 Multiple epidermal growth factor-like domains protein 2 RESDA1; cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila); EGF-like protein 1; EGF-like-domain, multiple 1; Epidermal growth factor-like protein 1; Flamingo homolog 1; hFmi1; Multiple EGF-like domains protein 2; Multiple epidermal growth factor-like domains protein 2
Gene Aliases: ADGRC3; CDHF11; CELSR3; EGFL1; FMI1; HFMI1; KIAA0812; MEGF2; RESDA1
UniProt ID: (Human) Q9NYQ7
Entrez Gene ID: (Human) 1951
Molecular Function: G-protein coupled receptor cadherin cell adhesion molecule receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.