Abnova
Manufacturer Code:PAB8720
Catalog # 89-133-878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 10 of mouse Fkbp1a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Fkbp1a.
For Research Use Only
Protein Aliases: 12 kDa FK506-binding protein; 12 kDa FKBP; 12kDa; Calstabin-1; FK506-binding protein 1A; Fkbp; FKBP-1A; Fkbp1; FKBP12; FKBP12-T1; FKBP12-T2; Immunophilin FKBP12; mFKBP1; mFKBP12; Peptidyl-prolyl cis-trans isomerase FKBP1A; PPIase FKBP1A; Rotamase
Gene Aliases: Fkbp; Fkbp1; FKBP12; Fkbp1a
UniProt ID: (Mouse) P26883
Entrez Gene ID: (Mouse) 14225
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.