Novus Biologicals
Manufacturer Code:NBP246572
Catalog # NBP246572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B2 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 405-665 of human BFSP1 (NP_001186) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beaded filament structural protein 1; Beaded filament structural protein 1 beaded filament structural protein 1 filensin CP115Lens intermediate filament-like heavy filensin Lens fiber cell beaded-filament structural protein CP 115 LIFL-H115 KD; beaded filament structural protein 1, filensin; CP115; cytoskeletal protein, 115 KD; Filensin; Filensin C-terminal fragment; Filensin N-terminal fragment; Lens fiber cell beaded-filament structural protein CP 115; Lens intermediate filament-like heavy; LIFL-H
Gene Aliases: BFSP1; CP115; CP94; CTRCT33; LIFL-H
UniProt ID: (Human) Q12934
Entrez Gene ID: (Human) 631
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.