Abnova
Manufacturer Code:MAB18664
Catalog # 89-338-589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S202-7 |
Immunogen | Recombinant protein corresponding to amino acids 688-907 at C-terminus of mouse Fig4. |
Conjugate | PE/ATTO594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A530089I17Rik; AI326867; Phosphatidylinositol 3,5-bisphosphate 5-phosphatase; Polyphosphoinositide phosphatase; Sac domain-containing inositol phosphatase 3; SAC domain-containing protein 3; Sac3
Gene Aliases: A530089I17Rik; AI326867; Fig4; Kiaa0274; Sac3
UniProt ID: (Mouse) Q91WF7
Entrez Gene ID: (Mouse) 103199
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.