Novus Biologicals
Manufacturer Code:NBP221613G
Catalog # NBP221613G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 16 |
Immunogen | Purified procaryot expressed recombinant human L-Ficolin and full-length L-Ficolin (Ficolin-2) protein adsorbed onto aluminum hydroxide gel |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 37 kDa elastin-binding protein; 37 kDa elastin-binding protein Collagen/fibrinogen domain-containing protein 2 EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin) FCNLficolin B ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin) ficolin-2 Ficolin-B Ficolin-beta Hucolin L-ficolin P35 Serum lectin p35; Collagen/fibrinogen domain-containing protein 2; EBP-37; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin B; Ficolin-2; Ficolin-B; Ficolin-beta; Hucolin; L-ficolin; Serum lectin p35
Gene Aliases: EBP-37; FCN2; FCNL; ficolin-2; P35
UniProt ID: (Human) Q15485
Entrez Gene ID: (Human) 2220
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.