Invitrogen
Manufacturer Code:MA515292
Catalog # PIMA515292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F10A5,3F8A12 |
Immunogen | Purified recombit fragment of Fibulin 5 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Dance; Developmental arteries and neural crest EGF-like protein; EVEC UP50 ARMD3 DANCE FIBL-5 FLJ90059 FBLN5; FIBL-5; Fibulin-5; testis tissue sperm-binding protein Li 75n; UP50; Urine p50 protein
Gene Aliases: ADCL2; ARCL1A; ARMD3; DANCE; EVEC; FBLN5; FIBL-5; HNARMD; UNQ184/PRO210; UP50
UniProt ID: (Human) Q9UBX5
Entrez Gene ID: (Human) 10516
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.