Invitrogen
Manufacturer Code:PA5109261
Catalog # PIPA5109261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein within human Fibulin 5 210-360 aa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A55 ADCL2 ARCL1A ARMD3 DANCE Developmental arteries and neural crest EGF-like protein Embryonic vascular EGF repeat-containing protein EVEC Fbln5 FIBL-5 fibulin 5 Fibulin-5 FLJ90059 HNARMD UNQ184/PRO210 UP50 Urine p50 protein; Dance; Developmental arteries and neural crest EGF-like protein; FIBL-5; Fibulin-5; testis tissue sperm-binding protein Li 75n; UP50; Urine p50 protein
Gene Aliases: ADCL2; ARCL1A; ARMD3; DANCE; EVEC; FBLN5; FIBL-5; HNARMD; UNQ184/PRO210; UP50
UniProt ID: (Human) Q9UBX5
Entrez Gene ID: (Human) 10516
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.