Proteintech
Manufacturer Code:121881AP150UL
Catalog # 50-172-7780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fibulin 5 Fusion Protein Ag2859 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARMD3 DANCE EVEC FBLN5 FIBL 5 fibulin 5 UP50 Urine p50 protein; Dance; Developmental arteries and neural crest EGF-like protein; Embryonic vascular EGF repeat-containing protein; EVEC; FIBL-5; Fibulin-5; testis tissue sperm-binding protein Li 75n; UP50; Urine p50 protein
Gene Aliases: A55; ADCL2; ARCL1A; ARMD3; DANCE; EVEC; FBLN5; FIBL-5; HNARMD; UNQ184/PRO210; UP50
UniProt ID: (Human) Q9UBX5, (Rat) Q9WVH8, (Mouse) Q9WVH9
Entrez Gene ID: (Human) 10516, (Rat) 29158, (Mouse) 23876
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.