Novus Biologicals
Manufacturer Code:NBP270700AF532
Catalog # NP27070A532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D9 |
Immunogen | Full length recombinant protein of human Fibulin-3/EFEMP1 (NP_004096) produced in HEK293T cells. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DHRD EGF containing fibulin-like extracellular matrix protein 1 EGF-containing fibulin-like extracellular matrix protein 1 Extracellular protein S1-5 FBLN3DRAD FBNLFLJ35535 FIBL-3 fibrillin-like Fibrillin-like protein Fibulin-3 MGC111353 MLVT MTLV S1-5; EGF containing fibulin-like extracellular matrix protein 1; EGF-containing fibulin-like extracellular matrix protein 1; Extracellular protein S1-5; FIBL-3; Fibrillin-like protein; Fibulin-3
Gene Aliases: DHRD; DRAD; EFEMP1; FBLN3; FBNL; FIBL-3; MLVT; MTLV; S1-5
UniProt ID: (Human) Q12805
Entrez Gene ID: (Human) 2202
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.