R&D Systems
Manufacturer Code:AF6235SP
Catalog # AF6235SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Fibulin 3 Gln19-Phe493 Accession # Q12805 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: DHRD EFEMP1 EGF containing fibulin-like extracellular matrix protein 1 EGF-containing fibulin-like extracellular matrix protein 1 Extracellular protein S1-5 FBLN3 FBLN3DRAD FBNLFLJ35535 FIBL-3 fibrillin-like Fibrillin-like protein Fibulin-3 MGC111353 MLVT MTLV S1-5; EGF containing fibulin-like extracellular matrix protein 1; EGF-containing fibulin-like extracellular matrix protein 1; Extracellular protein S1-5; FIBL-3; Fibrillin-like protein; Fibulin-3
Gene Aliases: DHRD; DRAD; EFEMP1; FBLN3; FBNL; FIBL-3; MLVT; MTLV; S1-5
UniProt ID: (Human) Q12805
Entrez Gene ID: (Human) 2202
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.